missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Nucleocapsid Phosphoprotein Rabbit Polyclonal Antibody, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 28769-1-AP-20UL

 View more versions of this product

Product Code. 30233206

  • € 145.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The SARS-CoV-2 Nucleocapsid Phosphoprotein antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of virus SARS-CoV-2 Nucleocapsid Phosphoprotein. This antibody recognizes virus antigen. The SARS-CoV-2 Nucleocapsid Phosphoprotein antibody has been validated for the following applications: WB, ELISA analysis.
Specifications

Specifications

SARS-CoV-2 Nucleocapsid Phosphoprotein
Polyclonal
Unconjugated
Recombinant Protein
20 μL
Primary
Virus
Polyclonal Antibody
IgG
Western Blot, ELISA
600 μg/mL
Rabbit
Antigen affinity purification
RUO
43740575
Store at -20°C. Stable for one year after shipment.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.