missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 ORF7a Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-11936MFV500
This item is not returnable.
View return policy
Description
SARS-CoV-2 ORF7a Polyclonal antibody specifically detects SARS-CoV-2 ORF7a in SARS-CoV-2 samples. It is validated for ELISA
Specifications
| SARS-CoV-2 ORF7a | |
| Polyclonal | |
| ELISA | |
| 2019-nCoV ORF7a, 2019-nCoV ORF7a Protein, Accessory Protein 7a, COVID-19 Accessory Protein 7a, COVID-19 ORF7a, COVID-19 Protein U122, COVID-19 Protein X4, Human coronavirus ORF7a Protein, ORF7a Protein, Protein U122, Protein X4, SARS-CoV-2 Accessory Protein 7a, SARS-CoV-2 Protein U122, SARS-CoV-2 Protein X4 | |
| Antibody was raised against a peptide corresponding to 9 amino acids near the carboxyl terminus of SARS-CoV-2 ORF7a protein. The immunogen is located within the last 50 amino acids of the SARS-CoV-2 ORF7a protein. | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 43740573 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| SARS-CoV-2 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction