missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-CoV-2 ORF9b Recombinant Rabbit Monoclonal Antibody (HL1918)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547221
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
ORF9b plays a role in the inhibition of host innate immune response by targeting the mitochondrial-associated adapter MAVS. Mechanistically, usurps the E3 ligase ITCH to trigger the degradation of MAVS, TRAF3, and TRAF6. In addition, causes mitochondrial elongation by triggering ubiquitination and proteasomal degradation of dynamin-like protein 1/DNM1L.
Specifications
| SARS-CoV-2 ORF9b | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL1918 | |
| Unconjugated | |
| COVID19 ORF9b; COVID-19 ORF9b; ORF9b; SARS-CoV-2 Open Reading Frame 9b; SARS-COV2 ORF9b | |
| Full length SARS-CoV-2 (COVID-19) ORF9b recombinant protein. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction