Learn More
IBA Lifesciences SARS-CoV-2 Peptide LALLLLDRL (HLA-A*0201)
Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
- SARS-CoV-2 peptide ILLNKHIDA (HLA-A*0201) is a single peptide for stimulation of T cells.
- The peptide from SARS-CoV-2 is synthesized as it is presented by the MHC class I HLA-A*0201 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
- The peptides were purified by HPLC using a water and acetonitrile gradient and are endotoxin-free.
Specifications
Specifications
| Purity | >85% |
| Endotoxin Level | Free |
| Format | Lyophilized Powder |
| Quantity | 1 mg |
| Research Category | T cell stimulation |
For Research Use Only.
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido