Learn More
IBA Lifesciences SARS-CoV-2 Peptide VLNDILSRL (HLA-A*0201)
GREENER_CHOICE
Product Code. 16644744 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. 16644744
Brand: IBA Lifesciences 67113901
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Single peptide synthesized as presented by the MHC class I allele
  • SARS-CoV-2 peptide VLNDILSRL (HLA-A*0201) is a single peptide for stimulation of T cells.
  • The peptide from SARS-CoV-2 is synthesized as it is presented by the MHC class I HLA-A*0201 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
  • The peptides were purified by HPLC using a water and acetonitrile gradient and are endotoxin-free.
Specifications
Purity >85%
Endotoxin Level Free
Format Lyophilized Powder
Quantity 1 mg
Research Category T cell stimulation