Learn More
IBA Lifesciences SARS-CoV-2 Peptide VVFLHVTYV (HLA-A*0201)
Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
- SARS-CoV-2 peptide VVFLHVTYV (HLA-A*0201) is a single peptide for stimulation of T cells.
- The peptide from SARS-CoV-2 is synthesized as it is presented by the MHC class I HLA-A*0201 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
- The peptides were purified by HPLC using a water and acetonitrile gradient and are endotoxin-free.
Specifications
Specifications
| Purity | >85% |
| Endotoxin Level | Free |
| Format | Lyophilized Powder |
| Quantity | 1 mg |
| Research Category | T cell stimulation |