missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 pp1ab polyprotein Antibody - Interior 1 - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05697
This item is not returnable.
View return policy
Description
SARS-CoV-2 pp1ab polyprotein Polyclonal antibody specifically detects SARS-CoV-2 pp1ab polyprotein in SARS-CoV-2 samples. It is validated for Western Blot, ELISA
Specifications
| SARS-CoV-2 pp1ab polyprotein | |
| Polyclonal | |
| Western Blot 1:500-1000, ELISA | |
| 2019-nCoV ORF1ab polyprotein, 2019-nCoV pp1ab, 2019-nCoV Replicase polyprotein 1ab, COVID-19 ORF1ab polyprotein, COVID-19 pp1ab, COVID-19 Replicase polyprotein 1ab, Human coronavirus Replicase polyprotein 1ab, ORF1ab polyprotein, pp1ab, Replicase polyprotein 1ab, SARS-CoV-2, SARS-CoV-2 ORF1ab polyprotein, SARSCoV2 pp1ab, SARS-CoV-2 Replicase polyprotein 1ab, Severe Acute Respiratory Syndrome Coronavirus 2 | |
| Synthetic peptide located within NSP3-IN1 of SARS-CoV-2 pp1ab polyprotein. | |
| 50 μg | |
| Infections (Virus Bacteria and Parasites) | |
| 43740578 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| PBS, 10% glycerol, pH7.2 | |
| Rabbit | |
| Epitope affinity purified | |
| RUO | |
| Primary | |
| SARS-CoV-2 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction