missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 pp1ab polyprotein Antibody [Janelia Fluor« 646], Novus Biologicals Biologicals™

Product Code. 30503631 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30503631 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30503631 Supplier Novus Biologicals Supplier No. NBP327904JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 pp1ab polyprotein Polyclonal antibody specifically detects SARS-CoV-2 pp1ab polyprotein in Virus samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 pp1ab polyprotein
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias 2019-nCoV ORF1ab polyprotein, 2019-nCoV pp1ab, 2019-nCoV Replicase polyprotein 1ab, COVID-19 ORF1ab polyprotein, COVID-19 pp1ab, COVID-19 Replicase polyprotein 1ab, Human coronavirus Replicase polyprotein 1ab, ORF1ab polyprotein, pp1ab, Replicase polyprotein 1ab, SARS-CoV-2, SARS-CoV-2 ORF1ab polyprotein, SARSCoV2 pp1ab, SARS-CoV-2 Replicase polyprotein 1ab, Severe Acute Respiratory Syndrome Coronavirus 2
Host Species Rabbit
Immunogen SARS-CoV-2 pp1ab polyprotein antibody was raised against a peptide corresponding to 16 amino acids near the central domain of SARS-CoV-2 pp1ab polyprotein protein. The immunogen is located within 120-170 amino acids of the SARS-CoV-2 pp1ab polyprotein protein.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 43740578
Target Species Virus
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.