missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Spike Human anti-SARS-CoV-2, Clone: D005, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP2-90989-100UL
This item is not returnable.
View return policy
Description
SARS Spike Protein Monoclonal specifically detects SARS Spike Protein in SARS-CoV-2, SARS-CoV samples. It is validated for ELISA.
Specifications
| SARS-CoV-2 Spike | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| SARS-CoV-2 | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| D005 | |
| Flow Cytometry, ELISA 1:5000 - 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, Human coronavirus spike glycoprotein, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein | |
| Recombinant SARS-CoV Spike RBD Protein | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction