Learn More
Invitrogen™ SARS-CoV-2 Spike Protein S1 NTD Recombinant Rabbit Monoclonal Antibody (235)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560385
Description
Recognize SARS-CoV-2 Spike S1-NTD. Doesn't recognize SARS-CoV-2 Spike RBD and S2. Has cross-reactivity with SARS-CoV.. Has cross-reactivity in ELISA and WB with: SARS-COV-2 Omicron (KP.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Spike S1+S2 ECD-His; SARS-CoV-2 Spike S1-His; SARS-CoV-2 Spike S1 NTD-Fc & AVI; SARS-CoV-2 Spike S1 NTD-His & AVI; SARS-CoV Spike S1+S2 ECD-His. Has cross-reactivity in ELISA with: SARS-CoV-2 (XEC) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB.1.16) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB.1.5) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB.1) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4/BA.5/BA.5.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BQ.1.1) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BF.7) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4.6) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.2.75) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.2.75.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (B.1.1.529) S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.1.1) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.2) Spike S1 Protein; SARS-CoV-2 Omicron (BA.2) Spike S1 NTD Protein; SARS-CoV-2 XD(BA.1 x AY.4) Spike S1+S2 trimer Protein; SARS-CoV-2 Delta (B.1.617.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Delta (B.1.617.2) Spike S1 Protein; SARS-CoV-2 Delta (B.1.617.2) Spike S1 NTD Protein; SARS-CoV-2 Spike S1 NTD-Fc & AVI; Medium cross-re...
SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. SARS-CoV-2 Spike Protein is composed of S1 domain and S2 domain (Spike Protein). The Spike protein is the main surface antigen of the coronavirus. The glycosylated Spike protein (as well as the nucleocapsid protein) can be detected in infected cell culture supernatants with antisera from SARS patients. S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2), the receptor on target cells. CoV diversity is reflected in the variable Spike proteins (S proteins), which have evolved into forms differing in their receptor interactions and their response to various environmental triggers of virus-cell membrane fusion.
Specifications
| SARS-CoV-2 Spike Protein S1 NTD | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 235 | |
| Unconjugated | |
| Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 | |
| Recombinant SARS-CoV-2 Spike S1 protein (Val16-Arg685). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.