missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-CoV-2 Spike Protein S1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120686
This item is not returnable.
View return policy
Description
Positive test controls include: 293T. Immunogen sequence: PGTNTSNQVA VLYQDVNCTE VPVAIHADQL TPTWRVYSTG SNVFQTRAGC LIGAEHVNNS YECDIPIGAG ICASYQTQTN SPRRARSVAS QSIIAYTMSL G.
SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. SARS-CoV-2 Spike Protein is composed of S1 domain and S2 domain (Spike Protein). The Spike protein is the main surface antigen of the coronavirus. The glycosylated Spike protein (as well as the nucleocapsid protein) can be detected in infected cell culture supernatants with antisera from SARS patients. S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2), the receptor on target cells. CoV diversity is reflected in the variable Spike proteins (S proteins), which have evolved into forms differing in their receptor interactions and their response to various environmental triggers of virus-cell membrane fusion.
Specifications
| SARS-CoV-2 Spike Protein S1 | |
| Polyclonal | |
| Unconjugated | |
| Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 | |
| A synthetic peptide corresponding to a sequence within amino acids 600-700 of coronavirus Spike (YP_0097243901). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Dot Blot, ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1.2 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction