Learn More
SARS-CoV-2 Spike RBD Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand:  R&D Systems MAB12504-100
 View more versions of this product
Product Code. 18199450
  • € 502.00 / 100µg
Description
Spike RBD Monoclonal antibody specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for Neutralization, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Spike RBD
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Rabbit
Protein A or G purified from cell culture supernatant
RUO
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution.
IgG
Neutralization, Immunocytochemistry
2820D
Neutralization, Immunocytochemistry 1-3 ug/mL
YP_009724390.1
Human embryonic kidney cell HEK293-derived SARS-CoV-2 Spike RBD Protein, Ala319-Phe541, Accession # YP_009724390.1
100 μg
Primary
SARS-CoV-2