Learn More
SARS-CoV-2 Spike S1 Protein Antibody (1035206), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB105403CL1
Product Code. 30148321
  • € 997.00 / 0.10mL
Description
SARS-CoV-2 Spike S1 Protein Monoclonal antibody specifically detects SARS-CoV-2 Spike S1 Protein in SARS-CoV-2 samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SARS-CoV-2 Spike S1 Protein
Monoclonal
CoraFluor 1
SARS-CoV-2
HEK293-derived SARS-CoV-2 Spike S1 Subunit protein, Accession # YP_009724390.1
0.1 mL
Primary
Store at 4°C in the dark. Do not freeze.
IgG1
Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF
1035206
PBS
Mouse
Protein A or G purified
RUO
SARS-CoV-2