missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Spike S1 Protein Antibody (1035226), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB105805PEATT594
This item is not returnable.
View return policy
Description
SARS-CoV-2 Spike S1 Protein Monoclonal antibody specifically detects SARS-CoV-2 Spike S1 Protein in SARS-CoV-2 samples. It is validated for Flow Cytometry
Specifications
| SARS-CoV-2 Spike S1 Protein | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| SARS-CoV-2 | |
| Purified |
| Flow Cytometry | |
| 1035226 | |
| Flow Cytometry | |
| SARS-CoV-2 | |
| HEK293-derived SARS-CoV-2 Spike S1 Subunit, Accession # YP_009724390.1 | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction