Learn More
R&D Systems™ SARS-CoV Nucleocapsid (YP_009825061.1) VersaClone cDNA
Click to view available options
:
10 μg
Spécification
Spécification
| Gene | Nucleocapsid |
| Accession Number | YP_009825061.1 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1282 bp |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | SARS-CoV |
| Quantity | 10 μg |