missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sars Envelope Protein Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 644.00
Specifications
| Antigen | Sars Envelope Protein |
|---|---|
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 450 SE |
| Form | Purified |
Description
Sars Envelope Protein Polyclonal antibody specifically detects Sars Envelope Protein in SARS-CoV samples. It is validated for ELISASpecifications
| Sars Envelope Protein | |
| Polyclonal | |
| Purified | |
| RUO | |
| SARS-CoV | |
| 1489671 | |
| IgG | |
| Peptide affinity purified |
| ELISA | |
| mFluor Violet 450 SE | |
| Rabbit | |
| Immunology, Virology Bacteria and Parasites | |
| SARS coronavirus Envelope protein, SARS CoV E protein, SARS CoV Envelope protein, SARSCoV, SARSCoV Envelope protein, Severe acute respiratory syndrome | |
| Anti-Sars Envelope Protein antibody was raised against a peptide corresponding to 11 amino acids near the carboxy terminus of SARS-CoV Envelope protein. The immunogen is located within the last 50 amino acids of SARS Envelope Proteins. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title