missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sars Membrane Protein Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41059B
This item is not returnable.
View return policy
Description
Sars Membrane Protein Polyclonal antibody specifically detects Sars Membrane Protein in SARS-CoV samples. It is validated for ELISA
Specifications
| Sars Membrane Protein | |
| Polyclonal | |
| ELISA | |
| E1 glycoprotein, Matrix glycoprotein, Membrane glycoprotein, SARS coronavirus M protein, SARS coronavirus Membrane protein, SARS CoV, SARS CoV M glycoprotein, SARS CoV Matrix protein, Sars M, Sars Matrix Protein, SARSCoV, SARS-CoV M protein, SARS-CoV Membrane protein, Severe acute respiratory syndrome | |
| The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to a 13 aa peptide from within the first 50 amino acids from the M (Membrane protein) for the human SARS coronavirus (Genbank accession no. AFR58746.1) Amino Acid Squence: ADNGTITVEELKQ | |
| 0.1 mL | |
| Immunology, Virology Bacteria and Parasites | |
| 1489672 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| Biotin | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| SARS-CoV | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction