missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS Nucleocapsid Protein Antibody (001), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90988H
This item is not returnable.
View return policy
Description
SARS Nucleocapsid Protein Monoclonal specifically detects SARS Nucleocapsid Protein in SARS-CoV-2, SARS-CoV samples. It is validated for Western Blot, ELISA.
Specifications
| SARS Nucleocapsid Protein | |
| Monoclonal | |
| HRP | |
| PBS | |
| N | |
| Recombinant SARS-CoV Nucleoprotein/NP Protein | |
| 0.1 mL | |
| Primary | |
| Has cross-reactivity in ELISA and WB with: SARS-CoV-2 Nucleocapsid Protein Has cross-reactivity in ELISA with: SARS-CoV-2 Omicron (B.1.1.529) Nucleocapsid Protein SARS-CoV-2 Nucleocapsid Protein SARS-CoV-2 Alpha(B.1.1.7) Nucleocapsid(R203K, G204R) Protein SARS-CoV-2 Alpha(B.1.1.7) Nucleocapsid(I292T) Protein SARS-CoV-2 Alpha(B.1.1.7) Nucleocapsid(D3L, R203K, G204R, S235F) Protein SARS-CoV-2 Alpha(B.1.1.7) Nucleocapsid(D3L, S235F) Protein SARS-CoV-2 Alpha(B.1.1.7/ B.1.237) Nucleocapsid(S194L) Protein SARS-CoV-2 Alpha/beta(B.1.1.7/ B.1.351/ A.2.2) Nucleocapsid(P13L) Protein SARS-CoV-2 Beta(B.1.351/B.1.351.2/b.1.351.3/B.1.427/B.1.429) Nucleocapsid (T205I) Protein SARS-CoV-2 Gamma(P.1/ P.1.1/ P.1.2) Nucleocapsid (P80R) Protein SARS-CoV-2 Eta(B.1.525) Nucleocapsid (A12G, T205I) Protein SARS-CoV-2 (B.1.617) Nucleocapsid (D377Y) Protein SARS-CoV-2 (B.1.617) Nucleocapsid (R203M, D377Y) Protein SARS-CoV-2 Delta(B.1.617.2) Nucleocapsid (D63G, R203M, D377Y) Protein SARS-CoV-2 (B.1.617.3) Nucleocapsid (P67S, R203M, D377Y) Protein No cross-reactivity in ELISA with: MERS-CoV Nucleocapsid protein HCoV-229E Nucleocapsid protein HCoV-NL63 Nucleocapsid protein HCoV-HKU1(isolate N5) Nucleocapsid protein HCoV-OC43 Nucleocapsid protein | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| 1 | |
| Western Blot, ELISA | |
| N, N protein, N structural protein, NC, Nucleocapsid protein, Nucleoprotein, Protein N, SARS coronavirus N protein, SARS coronavirus nucleocapsid protein, SARS CoV, SARS CoV N protein, SARS CoV nucleocapsid protein, SARS N protein, SARS Nucleoprotein, SARSCoV, SARSCoV N protein, SARSCoV nucleocapsid protein, Severe acute respiratory syndrome | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 1489678 | |
| SARS-CoV-2, SARS-CoV | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction