missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS Nucleocapsid Protein Antibody (05), Alexa Fluor™ 532 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06413AF532
This item is not returnable.
View return policy
Description
SARS Nucleocapsid Protein Monoclonal antibody specifically detects SARS Nucleocapsid Protein in SARS-CoV samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SARS Nucleocapsid Protein | |
| Monoclonal | |
| Alexa Fluor 532 | |
| N, N protein, N structural protein, NC, Nucleocapsid protein, Nucleoprotein, Protein N, SARS coronavirus N protein, SARS coronavirus nucleocapsid protein, SARS CoV, SARS CoV N protein, SARS CoV nucleocapsid protein, SARS N protein, SARS Nucleoprotein, SARSCoV, SARSCoV N protein, SARSCoV nucleocapsid protein, Severe acute respiratory syndrome | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant SARS Nucleocapsid Protein (NP_828858.1; Met1-Ala422). | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 1489678 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 05 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| SARS-CoV | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction