missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS Nucleocapsid Protein Antibody (05), Novus Biologicals™
Mouse Monoclonal Antibody
€ 252.00 - € 448.00
Specifications
| Antigen | SARS Nucleocapsid Protein |
|---|---|
| Clone | 05 |
| Dilution | Western Blot 1:1000-1:10000, Flow Cytometry 1:25-1:100, ELISA 1:1000-1:2000, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18610597
|
Novus Biologicals
NBP3-06413-100ul |
100 μg |
€ 448.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18629348
|
Novus Biologicals
NBP3-06413-50ul |
50 μg |
€ 252.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SARS Nucleocapsid Protein Monoclonal antibody specifically detects SARS Nucleocapsid Protein in SARS-CoV samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry (Paraffin)Specifications
| SARS Nucleocapsid Protein | |
| Western Blot 1:1000-1:10000, Flow Cytometry 1:25-1:100, ELISA 1:1000-1:2000, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| SARS-CoV | |
| N, N protein, N structural protein, NC, Nucleocapsid protein, Nucleoprotein, Protein N, SARS coronavirus N protein, SARS coronavirus nucleocapsid protein, SARS CoV, SARS CoV N protein, SARS CoV nucleocapsid protein, SARS N protein, SARS Nucleoprotein, SARSCoV, SARSCoV N protein, SARSCoV nucleocapsid protein, Severe acute respiratory syndrome | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant SARS Nucleocapsid Protein (Accession#: NP_828858.1; Met1-Ala422). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 05 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Infections (Virus Bacteria and Parasites) | |
| 0.2 um filtered solution in PBS | |
| 1489678 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title