missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS RDRP Antibody (4E6), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50258B
This item is not returnable.
View return policy
Description
SARS RDRP Monoclonal antibody specifically detects SARS RDRP in SARS-CoV samples. It is validated for Western Blot
Specifications
| SARS RDRP | |
| Monoclonal | |
| Biotin | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot | |
| 4E6 | |
| PBS | |
| The antibody was developed by immunizing mice with a GST fusion protein corresponding to Replicase polyprotein 1ab for the Human SARS coronavirus (Genbank accession no. NP_828849.2) | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| SARS-CoV | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction