missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS RDRP Antibody (4E6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 190.00 - € 361.00
Specifications
| Antigen | SARS RDRP |
|---|---|
| Clone | 4E6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000 |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18645728
|
Novus Biologicals
NBP2-50258-0.025ml |
0.025 mL |
€ 190.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671139
|
Novus Biologicals
NBP2-50258 |
0.1 mL |
€ 361.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SARS RDRP Monoclonal antibody specifically detects SARS RDRP in SARS-CoV samples. It is validated for Western BlotSpecifications
| SARS RDRP | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Virology Bacteria and Parasites | |
| PBS | |
| IgG1 | |
| Protein A purified |
| 4E6 | |
| Western Blot 1:100 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| SARS-CoV | |
| The antibody was developed by immunizing mice with a GST fusion protein corresponding to Replicase polyprotein 1ab for the Human SARS coronavirus (Genbank accession no. NP_828849.2) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title