missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS Spike Protein Antibody (16F1071), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-24746C
This item is not returnable.
View return policy
Description
SARS Spike Protein Monoclonal specifically detects SARS Spike Protein in SARS-CoV samples. It is validated for Western Blot.
Specifications
| Sars Spike Protein | |
| Monoclonal | |
| DyLight 650 | |
| P49591 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| The C1.18.4 hybridoma cell line was grown and tissue culture supernatant was purified using affinity chromatography. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot | |
| 16F1071 | |
| Western Blot | |
| E2, E2 glycoprotein, Human coronavirus spike glycoprotein, Peplomer protein, S, S glycoprotein, Severe acute respiratory syndrome spike glycoprotein, Severe acute respiratory syndrome virus spike glycoprotein, Spike glycoprotein, VGL2 | |
| The antibody was developed using a synthetic peptide from the putative SARS Spike glycoprotein (Genbank accession no. NP_828851) corresponding to amino acids 19-45. | |
| 0.1 mL | |
| Immunology, Virology Bacteria and Parasites | |
| 1489668 | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction