missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS Spike Protein Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56578AF532
This item is not returnable.
View return policy
Description
SARS Spike Protein Polyclonal antibody specifically detects SARS Spike Protein in SARS-CoV, SARS-CoV-2 samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| SARS Spike Protein | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Alexa Fluor 532 | |
| E2, E2 glycoprotein, Human coronavirus spike glycoprotein, Peplomer protein, S, S glycoprotein, SARS coronavirus S protein, SARS coronavirus Spike Protein, SARS CoV, SARS CoV S glycoprotein, SARS CoV Spike protein, SARSCoV, SARS-CoV S protein, SARS-COV Spike, Severe acute respiratory syndrome spike glycoprotein, Severe acute respiratory syndrome virus spike glycoprotein, Spike glycoprotein, VGL2 | |
| The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 1124-1140 (C-GNCDVVIGIVNNTVYDP) from the S (Spike glycoprotein)(Spike protein S2') for the Human SARS coronavirus (Genbank accession no. YP_009724390) | |
| 0.1 mL | |
| Core ESC Like Genes, Immunology, Stem Cell Markers, Virology Bacteria and Parasites | |
| SARS-CoV, SARS-CoV-2 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction