missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS Spike Protein Antibody (D005), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90989CL1
This item is not returnable.
View return policy
Description
SARS Spike Protein Monoclonal antibody specifically detects SARS Spike Protein in SARS-CoV,SARS-CoV-2 samples. It is validated for ELISA
Specifications
| SARS Spike Protein | |
| Monoclonal | |
| CoraFluor 1 | |
| E2, E2 glycoprotein, Human coronavirus spike glycoprotein, Peplomer protein, S, S glycoprotein, SARS coronavirus S protein, SARS coronavirus Spike Protein, SARS CoV, SARS CoV S glycoprotein, SARS CoV Spike protein, SARSCoV, SARS-CoV S protein, SARS-COV Spike, Severe acute respiratory syndrome spike glycoprotein, Severe acute respiratory syndrome virus spike glycoprotein, Spike glycoprotein, VGL2 | |
| Recombinant SARS-CoV Spike RBD Protein | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| ELISA | |
| D005 | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| SARS-CoV, SARS-CoV-2 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction