missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SART3, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00009733-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MATAAETSASEPEAESKAGPKADGEEDEVKAARTRRKVLSRAVAAATYKTMGPAWDQQEEGVSESDGDEYAMASSAESSPGEYEWEYDEEEEKNQLEIERLEEQVGPGVGSGHLPVFQVLGSPCPGPPPSpecifications
| SART3 | |
| Polyclonal | |
| No additive | |
| BC041638.1 | |
| SART3 | |
| SART3 (AAH41638.1, 1 a.a. ∼ 129 a.a) full-length human protein. | |
| RUO | |
| 9733 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| squamous cell carcinoma antigen recognized by T cells 3 | |
| DSAP1/KIAA0156/MGC138188/P100/RP11-13G14/TIP110/p110/p110(nrb) | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction