Learn More
Sartorius™ Sartobran™ Capsules
Used for sterile filtration of blood and plasma products and vaccines.
Brand: Sartorius™ 5231307H5--OO--B
Additional Details : Weight : 0.44100kg
Description
- With low-binding cellulose acetate membrane
- High product yield due to ultra-low unspecific adsorption
- Filtration results especially for large, shear-stress sensitive proteins
Specifications
Autoclavable | |
0.2 μm | |
5 Pack |
Cellulose Acetate | |
Sterile | |
Filter Capsule |