missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- The Sartolab™ BT bottle top filter is comprised of a graduated funnel with a polyethersulfone (PES) membrane
- A vacuum adapter and a lid, a tube connector for vacuum connection (for stand-alone filtration)
Specifications
Specifications
| Disposable | Single-use |
| For Use With (Equipment) | Bottle |
| Material | Polystyrene |
| Pore Size | 0.22 μm |
| Sample Volume | 0.15 μL |
| Sterility | Sterile |
| Type | Bottle Top Filter |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?