Learn More
Sartorius Absorbent Pads
Shop All Sartorius Products
Click to view available options
Packaging Type:
Packaged in 10 magazines
Packaged in petridish
Diameter (English) Outside:
1.85 in.
1.96 in.
Diameter (Metric) Outside:
4.7 cm
5 cm
50 mm
Unit Size:
Pack of 100
Pack of 1000
Description
Sartorius 1.4mm thick absorbent pads are wetted with the appropriate liquid culture medium before a membrane filter is placed on them. They come presterilized in plastic magazines, which fit onto the Sartorius manual dispensing device.
The absorbent pads are available in two diameters:
- 47mm has approx. 3ml absorption capacity
- 50mm with approx. 3.5ml absorption capacity
Specifications
Specifications
| Product Type | Nutrient Pad |
| Packaging Type | Packaged in 10 magazines |
| Diameter (English) Outside | 1.85 in. |
| Diameter (Metric) Outside | 4.7 cm |