Learn More
Sartorius Accessories for Combisart™ Systems: Glass Funnel Adapters
Shop All Sartorius Products
Click to view available options
Holds:
250mL glass funnel
30mL glass funnel
Unit Size:
Each
Description
Glass funnel adapters for use with Sartorius Combisart Systems
- Stainless steel
- Silicone stoppers
Specifications
Specifications
| Product Type | Adapter |
| Display Type | None |
| Holds | 30mL glass funnel |
| Material | Aluminum |
| For Use With (Equipment) | Combisart Manifold |
| Includes | 11mm Opening in Silicone stopper |