Learn More
Sartorius Analytical Draft Shield for Quintix™ and Practum™ Toploader Balances
GREENER_CHOICE
Product Code. 15347516 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Quintix or Practum Balance
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15347516 Quintix or Practum Balance Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15347516 Supplier Sartorius Supplier No. YDS01SQP
Add to basket
Add to basket
Analytical draft ring.
Applicable for Quintix or Practum balances with a readability of 10mg and a weighing range of up to 2100g or Quintix/Practum balances with a readability of 0.1g or 1g.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Quintix or Practum Balance
Type Draft Ring