Learn More
Sartorius Cubis™ Essential Analytical Balances
The right choice for users without complex operations who primarily want to perform ultra-precise weighing
Brand:  Sartorius MSE324S-ED15
Additional Details : Weight : 12.90000kg
 View more versions of this product
Product Code. 15354278
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Features
  • Large, high-contrast, liquid crystal Cubis MSE display
  • Easy-to-understand menu guidance with short text prompts
  • Clearly structured keys for precise activation of functions
  • Software support for fast leveling
  • Robust, monolithic weighing system technology for reliable measuring results
  • Easy calibration with isoCAL (internal automatic calibration | adjustment function)
  • Amply dimensioned, rectangular weighing pans
  • Data Interfaces: USB and RS232C; 3 optional ports are available as an accessory (Bluetooth, PS/2, RS232C)
  • Choose from 13 preconfigured premium Cubis Lab Balances, including semi-micro, analytical, precision and high-capacity balances
  •  
Draft Shields
  • Semi-micro and analytical models (14-559-000, 001, 002)  include draft shields with high mechanical stability and effortlessly silent moving doors. 
  • No disturbing frame, for unhindered access to the weighing pan.
  • Protected against electrostatic influences by a conductive coating
Applications
Unit conversion, isoCAL automatic calibration/adjustment function, density determination (buoyancy method only), calculations, averaging, net/total formulation, weighing in percent, counting,
totalizing
Specifications
Specifications
0.1 mg
MSE324S-ED15
≤3 sec.
85 mm
≤0.3 mg
≤0.1 mg
Analytical Balance