Learn More
Sartorius Microsart™ Mini Vacuum Pumps
Product Code. 15952540 Shop All Sartorius Products
Click to view available options
Flow Rate:
6 L/min.
Unit Size:
Each
Product Code. 15952540
Brand: Sartorius 1669416006
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Microsart Mini vacuum pumps provide up-to-date technology for daily use in the microbiology laboratory environment
Ideal for microbiological filtrations
  • Neoprene membrane pumps
  • Decreased sub lethals, higher recovery rates and shorter incubation times
  • Low noise level
  • Oil-and maintenance-free
  • Controlled and easily adjustable vacuum
  • Pressure filtration with Microsart mini vac possible
TRUSTED_SUSTAINABILITY
Specifications
Max. Vacuum 100 mbar
Shipping Weight (English) 4.18 lb.
Shipping Weight (Metric) 1.9 kg
Amperage 0.63 A
Electrical Requirements 115 V 60 Hz
Dimensions (L x W x H) 6.5 x 3.5 x 5.6 in. (164 x 90 x 141 mm)
Noise Level 53.5 dB (A)
Wattage 65 w
Temperature (English) Max. 104°F
Temperature (Metric) Max 40°C
Flow Rate 6 L/min.
Frequency 60 Hz
Overall Height (English) 5.6 in.
Overall Height (Metric) 14.2 cm
Overall Length (English) 6.5 in.
Overall Length (Metric) 16.4 cm
Overall Width (English) 3.5 in.
Overall Width (Metric) 90 mm
Pump Type Vacuum Pump
Maximum Pressure 2.5 bar
Voltage 115 V