Learn More
Sartorius Pump Heads for Vivaflow™ Masterflex Pump Models
Shop All Sartorius Products
Click to view available options
For Use With:
Vivaflow™ 200
Vivaflow™ 50 and 200
Unit Size:
Each
Description
- Masterflex, Easy-Load models
- Requires Masterflex Pump VFP002
Specifications
Specifications
| For Use With | Vivaflow™ 50 and 200 |
| Type | Pump Head |