Learn More
Sartorius Sterility Testing Reusable System: Tubing Clamp
Product Code. 10072431 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Reusable Sterility Testing System
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10072431 Reusable Sterility Testing System Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10072431 Supplier Sartorius Supplier No. 16978
Tubing Clamp for use with Reusable Sterility Testing System.
  • Clamps off tubing between T or X adapter and cap
  • Use while culture medium is filled in a filter holder that is not clamped off
  • Not autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Disposable Reusable
For Use With (Equipment) Reusable Sterility Testing System
Material Stainless Steel
Sterility Non-sterile
Type Tube Clamps