Learn More
Sartorius Vivapure™ LentiSELECT 500 Purification and Concentration Kit
Shop All Sartorius Products
Description
Vivapure™ LentiSELECT 500 is optimally suited for VSV-G pseudotyped lentivirus purification from up to 500mL cell culture and contains all reagents and devices necessary for purifying up to 2-5 x 109 infective particles.
The whole purification procedure is simply operated by a laboratory pump, which min imizes hands-on time. Unlike conventional purification methods as ultracentrifugation, Vivapure LentiSELECT 500 offers a fast and simple solution for purifying VSV-G pseudotyped lentiviruses making expensive purification equipment like ultracentrifuges redundant.
Typically, for a normal yielding vector, a 500mL cell culture purified using this method will yield up to 2-5 x 109 infective particles in 1mL (total volume 1mL).
- Fast
- Non-toxic
- Better purity than pelleting by ultracentrifugation
- Convienient
- Cost effective
Specifications
Specifications
| Sample Size | 500 mL of Lentivirus |
| Purification Target | 40 to 1000 mL Cell Cultures |
| For Use With (Application) | VSV-G Pseudotyped Lentiviral Vectors |