missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SASH1 Monoclonal Antibody (X1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524573
This item is not returnable.
View return policy
Description
Peptide Sequence: ENTSLQEHGV KLGPALTRKV SCARGVDLET LTENKLHAEG IDLTEEPYSD KHGRCGIPEA LVQRYAEDLD QPERDVAANM DQIRVKQLRK QHRMAIPSGG LTEICRKPVS.
May have a role in a signaling pathway. Could act as a tumor suppressor.
Specifications
| SASH1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| O94885 | |
| SASH1 | |
| SASH1 (NP_056093, 1066 a.a. ~ 1175 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| X1 | |
| Unconjugated | |
| SASH1 | |
| 1100001C18Rik; 2500002E12Rik; A330076K04Rik; dJ323M4; dJ323M4.1; KIAA0790; mKIAA0790; PEPE1; Proline-glutamate repeat-containing protein; putative adapter and scaffold protein 1; RGD1566017; SAM and SH3 domain containing 1; SAM and SH3 domain-containing protein 1; Sash1; SH3D6A | |
| Mouse | |
| Protein A | |
| RUO | |
| 23328 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction