missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SASS6 Antibody [PerCP], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NB100-93342PCP
This item is not returnable.
View return policy
Description
SASS6 Polyclonal antibody specifically detects SASS6 in Human samples. It is validated for Immunoprecipitation,Western Blot (Negative)
Specifications
| SASS6 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot (Negative) | |
| PerCP | |
| DKFZp761A078, FLJ22097, HsSAS-6, MGC119440, SAS-6, SAS6hsSAS-6, spindle assembly 6 homolog (C. elegans), spindle assembly abnormal protein 6 homolog | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human spindle assembly 6 using the numbering given in entry NP_919268.1 (GeneID 163786). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 163786 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction