missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAT2 Antibody (OTI1C9), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73990PEATT594
This item is not returnable.
View return policy
Description
SAT2 Monoclonal specifically detects SAT2 in Human samples. It is validated for Flow Cytometry.
Specifications
| SAT2 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| SAT2 | |
| Full length recombinant protein of human SAT2 (NP_597998) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| OTI1C9 | |
| Flow Cytometry | |
| diamine N-acetyltransferase 2, EC 2.3.1.57, Polyamine N-acetyltransferase 2, S, Spermidine/spermine N(1)-acetyltransferase 2, spermidine/spermine N1-acetyltransferase 2, spermidine/spermine N1-acetyltransferase family member 2, SSAT2diamine acetyltransferase 2, Thialysine N-epsilon-acetyltransferase | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 112483.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction