missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAT2, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00112483-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MASVRIREAKEGDCGDILRLIRELAEFEKLSDQVKISEEALRADGFGDNPFYHCLVAEILPAPGKLLGPCVVGYGIYYFIYSTWKGRTIYLEDIYVMPEYRGQGIGSKIIKKVAEVALDKGCSQFRLAVLDWNQRAMDLYKALGAQDLTEAEGWHFFCFQGEATRKLAGKSpecifications
| SAT2 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_133491.2 | |
| SAT2 | |
| SAT2 (NP_597998.1, 1 a.a. ∼ 170 a.a) full-length human protein. | |
| RUO | |
| 112483 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| spermidine/spermine N1-acetyltransferase family member 2 | |
| SSAT2 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction