missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SATB1 Antibody (OTI13D6), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73991FR
This item is not returnable.
View return policy
Description
SATB1 Monoclonal specifically detects SATB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| SATB1 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| SATB1 | |
| Full-length protein expressed in 293T cell transfected with human SATB1 expression vector. | |
| 0.1 mL | |
| 6304.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI13D6 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DNA-binding protein SATB1, SATB homeobox 1, special AT-rich sequence binding protein 1 (binds to nuclearmatrix/scaffold-associating DNA′s), Special AT-rich sequence-binding protein 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction