missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SATB1 Antibody (SATB1/2661) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27006AF350
This item is not returnable.
View return policy
Description
SATB1 Monoclonal antibody specifically detects SATB1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SATB1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| DNA-binding protein SATB1, SATB homeobox 1, special AT-rich sequence binding protein 1 (binds to nuclearmatrix/scaffold-associating DNA's), Special AT-rich sequence-binding protein 1 | |
| Recombinant full-length human SATB1 protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| SATB1/2661 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 6304 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction