missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SATB2 Antibody (SATB2/8292R), Alexa Fluor™ 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24251AF594
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
SATB2 Monoclonal antibody specifically detects SATB2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Tekniske data
| SATB2 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| DNA-binding protein SATB2, FLJ32076, KIAA1034FLJ21474, MGC119474, MGC119477, SATB family member 2, SATB homeobox 2, Special AT-rich sequence-binding protein 2 | |
| Recombinant fragment (around aa150-350) of human SATB2 (exact sequence is proprietary) | |
| 0.1 mL | |
| Signal Transduction | |
| 23314 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| SATB2/8292R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion