missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SATB2 Monoclonal Antibody (CL0276)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531339
This item is not returnable.
View return policy
Description
Immunogen sequence: VSQAVFARVA FNRTQGLLSE ILRKEEDPRT ASQSLLVNLR AMQNFLNLPE VERDRIYQDE RERSMNPNVS MVSSASSSPS SSRTPQAKTS TPTTDLPIKV DGANINITAA IYDEIQQEMK RAK Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.
DNA-binding protein SATB2 binds to DNA at the nuclear matrix- or scaffold associated regions. It is thought to recognize the sugar-phosphate structure of double-stranded DNA. SATB2 is a transcription factor controlling nuclear gene expression by binding to matrix attachment regions (MARs) of DNA and inducing a local chromatin-loop remodeling. It also acts as a docking site for several chromatin remodeling enzymes by recruiting corepressors (HDACs) or coactivators (HATs) directly to promoters and enhancers. SATB2 is required for the initiation of the upper-layer neurons (UL1) specific genetic program and for the inactivation of deep-layer neurons (DL) and UL2 specific genes - probably by modulating BCL11B expression. It is a repressor of Ctip2 and a regulatory determinant of corticocortical connections in the developing cerebral cortex. Mutations in the gene are associate with cleft palate isolated (CPI).
Specifications
| SATB2 | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UPW6 | |
| SATB2 | |
| Recombinant protein fragment of Human SATB2 (Product #RP-93680) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| CL0276 | |
| Unconjugated | |
| SATB2 | |
| DNA-binding protein SATB2; FLJ21474; FLJ32076; GLSS; KIAA1034; KIAA1034-like DNA binding protein; MGC119474; MGC119477; mKIAA1034; RGD1562369; SATB family member 2; SATB homeobox 2; SATB2; special AT-rich sequence binding protein 2; special AT-rich sequence-binding protein 2; two cut domains-containing homeodomain protein | |
| Mouse | |
| Protein A | |
| RUO | |
| 23314 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction