missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Satumomab Chimeric Recombinant Rabbit Monoclonal Antibody (B72.3 (Satumomab))

Description
Specificity: Recognises human Tumor associated glycoprotein 72.
Satumomab is a biosimilar that targets TAG-72.
Specifications
Specifications
| Antigen | Satumomab Chimeric |
| Applications | Flow Cytometry, Immunohistochemistry, Immunomicroscopy |
| Classification | Recombinant Monoclonal |
| Clone | B72.3 (Satumomab) |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene Alias | Indium (111In) satumomab pendetide; OncoScint CR; OncoScint CR103 |
| Host Species | Rabbit |
| Immunogen | human Tumor associated glycoprotein 72. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?