Learn More
Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Product Code. 10399102 Shop All Thermo Scientific Products
Click to view available options
Type:
Acid Neutralization Cartridge
Acid Neutralizing Compound
Ammonia Neutralizing Solution
Cart
Chemical Trap Kit
Condensation Flask
Disposable Cartridge
Heat Transfer Fluid
Unit Size:
0.25kg
5 L
Each
Pack of 4
Product Code. 10399102
Brand: Thermo Scientific™ ANS121
Improve the functionality of Savant SpeedVac Concentrators with these Savant SpeedVac Concentrators miscellaneous accessories.
Improve the funcionality of Thermo Scientific™ Savant™ SpeedVac™ Concentrators with Savant™ SpeedVac™ Concentrators Accessories.
TRUSTED_SUSTAINABILITY
Specifications
Description Ammonia Neutralizing Solution (Oligo Prep)
Type Ammonia Neutralizing Solution
For Use With (Equipment) All SpeedVac™ Unit
Model ANS121
Quantity Each
compliance-icons compliance-icons
Product Identifier
  • Neutralizing Solution
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P331-Do NOT induce vomiting.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337-If eye irritation persists:.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : citric acid monohydrate, dimethylpolysiloxane, D-limonene, thymol blue