missing translation for 'onlineSavingsMsg'
Learn More

SB 271046 Hydrochloride, MedChemExpress

Product Code. 16422177
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16422177

Brand: Medchem Express HY14336A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SB 271046 Hydrochloride (SB 271046A) is a potent, selective and orally active 5-HT6 receptor antagonist with pKi of 9.02, 8.55, and 8.81 for rat, pig and human, respectively. SB 271046 Hydrochloride is over 200 fold selective for the 5-HT6 receptor vs 55 other receptors, binding sites and ion channels. Anticonvulsant activity (EC50=0.16 μM).

SB 271046 Hydrochloride (SB 271046A) is a potent, selective and orally active 5-HT6 receptor antagonist with pKi of 9.02, 8.55, and 8.81 for rat, pig and human, respectively. SB 271046 Hydrochloride is over 200 fold selective for the 5-HT6 receptor vs 55 other receptors, binding sites and ion channels. Anticonvulsant activity (EC50=0.16 μM).

Specifications

Chemical Name or Material SB 271046 Hydrochloride
CAS 209481-24-3
Color White
Purity Grade Notes Research
Molecular Formula C20H23Cl2N3O3S2
Quantity 50 mg
Synonym SB 271046A
Solubility Information DMSO : 50 mg/mL (102.36 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.05 mM; ultrasonic and warming and heat to 60°C)
SMILES ClC1=CC2=C(C=C1)SC(S(=O)(NC3=CC=C(C(N4CCNCC4)=C3)OC)=O)=C2C.[H]Cl
Molecular Weight (g/mol) 488.45
Formula Weight 488.45
Percent Purity 98.64%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.