missing translation for 'onlineSavingsMsg'
Learn More

SB-334867 free base, MedChemExpress

Product Code. 16400587
Encompass
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16400587

Brand: Medchem Express HY10895A

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress SB-334867 free base (SB334867A free base) is an excellent, selective and blood-brain barrier permeable orexin-1 (OX1) receptor antagonist, shows selectivity over OX2 (pKb=7.4), 100-fold over 5-HT2B, 5-HT2C with pKi values of 5.4 and 5.3, respectively. SB-334867 reduces ethanol consumption and inhibits the acquisition of morphine-induced sensitization to locomotor activity in vivo.

SB-334867 free base (SB334867A free base) is an excellent, selective and blood-brain barrier permeable orexin-1 (OX1) receptor antagonist, shows selectivity over OX2 (pKb=7.4), 100-fold over 5-HT2B, 5-HT2C with pKi values of 5.4 and 5.3, respectively. SB-334867 reduces ethanol consumption and inhibits the acquisition of morphine-induced sensitization to locomotor activity in vivo.

Specifications

Chemical Name or Material SB-334867 free base
CAS 792173-99-0
Color White
Purity Grade Notes Research
Molecular Formula C17H13N5O2
Quantity 50 mg
Synonym SB334867A free base
Solubility Information DMSO : 50 mg/mL (156.58 mM; Need ultrasonic) ∣0.1 M HCL : 6 mg/mL (18.79 mM; ultrasonic and adjust pH to 3 with HCl)
SMILES O=C(NC1=CC(OC(C)=N2)=C2C=C1)NC3=CC=NC4=CC=CN=C43
Molecular Weight (g/mol) 319.32
Formula Weight 319.32
Percent Purity 99.33%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.