missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SB 525334
GREENER_CHOICE

Product Code. p-5546013
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18024892

Brand: Tocris Bioscience™ 3211/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective inhibitor of TGF-βRI

SB 525334 is a selective inhibitor of transforming growth factor-β receptor I (ALK5, TGF-βRI) (IC50 = 14.3 nM). Inhibits TGF-β1-induced smad2/3 nuclear localization and TGF-βRI-induced mRNA expression in kidney cells. Attenuates bleomycin-induced pulmonary fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type SB 525334
Purity >97%
Molecular Weight (g/mol) 343.42
Inhibitors SB 525334

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.