missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SBCAD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112303
This item is not returnable.
View return policy
Description
SBCAD Polyclonal Antibody for Western Blot, IHC (P), IP
Short/branched chain acyl-CoA dehydrogenase(ACADSB) is a member of the acyl-CoA dehydrogenase family of enzymes that catalyze the dehydrogenation of acyl-CoA derivatives in the metabolism of fatty acids or branch chained amino acids. Substrate specificity is the primary characteristic used to define members of this gene family. The ACADSB gene product has the greatest activity towards the short branched chain acyl-CoA derivative, (S)-2-methylbutyryl-CoA, but also reacts significantly with other 2-methyl branched chain substrates and with short straight chain acyl-CoAs. The cDNA encodes for a mitochondrial precursor protein which is cleaved upon mitochondrial import and predicted to yield a mature peptide of approximately 43.7- kDa.
Specifications
| SBCAD | |
| Polyclonal | |
| Unconjugated | |
| ACADSB | |
| 1300003O09Rik; 2-MEBCAD; 2-methyl branched chain acyl-CoA dehydrogenase; 2-methylbutyryl-CoA dehydrogenase; 2-methylbutyryl-coenzyme A dehydrogenase; ACAD7; Acadsb; acyl-CoA dehydrogenase short/branched chain; acyl-CoA dehydrogenase, short/branched chain; Acyl-Coenzyme A dehydrogenase short-branched chain; acyl-Coenzyme A dehydrogenase, short/branched chain; BB066609; SBCAD; Short/branched chain specific acyl-CoA dehydrogenase, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 36 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| P45954 | |
| ACADSB | |
| E. coli-derived Human ACADSB fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction